Learn More
DWK Life Sciences Kimble™ Kontes™ KIMAX™ NMR Borosilicate Glass 5mm Tube
Ideal for sealing directly to vacuum manifolds, joints or valves
Brand:  DWK Life Sciences 897241-0000
 View more versions of this product
Product Code. 10101352
  • € 183.00 / Pack of 5
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Description
  • Made of KIMAX KG-33 borosilicate glass
  • Have attached polyethylene cap
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Yes
177.8 mm
Polyethylene
0.0005 in.
0.00025 in.
600 to 700MHz
Cap
0.014 in.
0.19 in.
Borosilicate Glass
5 mm
0.16 in.
4.2 mm
Ideal for sealing directly to vacuum manifolds, joints or valves
7 in.
0.38 mm