Learn More
DWK Life Sciences Kimble™ Kontes™ Brand 5mm High Grade NMR Sample Tubes
Each tube meets rigid specifications for wall thickness, variation and camber so as to assure precise measurements and reproducibility.
Brand:  DWK Life Sciences 897200-0008
 View more versions of this product
Product Code. 10049701
  • € 43.85 / Pack of 5
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Description
Precision-manufactured tubes designed for use in high-resolution NMR spectroscopy that characterizes organic molecules through measurement of the resonant frequencies of atomic nuclei. Research applications include analysis of polymer structures and organic solutions.
  • Made from Type I borosilicate glass
  • Wall thickness: 0.01475 in. (0.37mm)
  • Wall variation and camber depend, in part, on tube length
  • All sample tubes come with plastic compression-fit caps and sandblasted marking spots
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.19 in.
Borosilicate Glass
Compression-fit
±0.003 in.
±0.002 in.
100MHz
Cap
Cylindrical
0.38 mm
203.2 mm
Polyethylene
5 mm
0.165 in.
4.2 mm
Ideal for sealing directly to vacuum manifolds, joints or valves
8 in.
0.014 in.