missing translation for 'onlineSavingsMsg'
Learn More

HMC Europe Vertical Autoclaves: HV Series

Programmable top-loading autoclaves with varying chamber volumes

Brand:  HMC Europe 20000

 View more versions of this product

Product Code. 10344391

  • € 8950.00 / Each
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

 

  • Use steam sterilization
  • Six sizes from 25 to 110 liters
  • Low height, easy-to-load
  • Portable — require no plumbing to operate but can be plumbed
  • Complete line of baskets, inserts and cans available separately
  • HV Series (dependable, hard-working, economical)
  • Many upgrades available depending on your needs

 

Certifications

CE marked

Services

Two years on parts; One year on labor

Specifications

Specifications

CE
95 cm
24 cm
25 L
21.65 in.
HV L25
105°C to 126°C
41 kg
46 cm
1.5 kw
48 cm
37.4 in.
9.44 in.
6.6 gal.
Steam sterilization
55 cm
221°F to 258.8°F
90.38 lb.
1 to 250 min.
18.11 in.
18.89 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.