Learn More
HMC Europe Vertical Autoclave Temperature Sensors
For the sterilization of liquids. Extremely flexible, mechanically and chemically resistant. Quick response, encapsulated, steam-tight.
Brand: HMC Europe 51001
This item is not returnable.
View return policy
Description
- Temperature sensing probe located inside the chamber.
- Designed so that the user can place the tip of the probe into the load being sterilized. It is usually placed in the most difficult to sterilize location.
Specifications
HMC-HG autoclave | |
Flexible Temperature Sensor |