Learn More
HMC Europe Vertical Autoclave Temperature Sensors
For the sterilization of liquids. Extremely flexible, mechanically and chemically resistant. Quick response, encapsulated, steam-tight.
Brand:  HMC Europe 51001
Product Code. 10772204
  • € 1076.83 / Each
Estimated Shipment: 10-03-2025
to see stock.
Description
  • Temperature sensing probe located inside the chamber.
  • Designed so that the user can place the tip of the probe into the load being sterilized. It is usually placed in the most difficult to sterilize location. 
Specifications
Specifications
HMC-HG autoclave
Flexible Temperature Sensor