Learn More
HMC Europe GLP Set for HV-Series Autoclaves
Combines two frequently ordered accessories: Printer and Exhaust Air Filter.
Brand:  HMC Europe 50021
Product Code. 10759410
  • € 5014.44 / Each
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Description
Set includes:
  • Printer for recording sterilization (data, temperature and time) 
  • Exhaust air filter (inclusive filter cartridge) for sterilizing high infectious materials
  • In-line-sterilization cartridge
Note:  Image shows the HV Autoclave family.
Specifications
Specifications
HMC Autoclaves, HV-Series
GLP Package
Exhaust Air Filter, Cartridge and Printer