Learn More
HMC Europe GLP Set for HV-Series Autoclaves
Combines two frequently ordered accessories: Printer and Exhaust Air Filter.
Brand: HMC Europe 50021
This item is not returnable.
View return policy
Description
Set includes:- Printer for recording sterilization (data, temperature and time)
- Exhaust air filter (inclusive filter cartridge) for sterilizing high infectious materials
- In-line-sterilization cartridge
Specifications
HMC Autoclaves, HV-Series | |
GLP Package | |
Exhaust Air Filter, Cartridge and Printer |