Learn More
DWK Life Sciences Kimble™ Kontes™ Potter-Elvehjem Tissue Grinders
Product Code. p-155723
Click to view available options
Capacity:
0.13 oz., 4mL
0.33 oz., 10mL
1.01 oz., 30mL
Graduations:
0.5 to 4mL
1 to 9mL
2 to 25mL
Unit Size:
Each
Product Code. 10604393
Brand: DWK Life Sciences 8855100020
Only null left
Add to basket
Add to basket
With serrated PTFE pestle and graduated tube
  • Radial serrations on the lower portion of PTFE pestle deliver homogenate into cylindrical portion of tube
  • Stainless-steel pestle shaft
  • Graduations fused into the glass tube
  • Tubes and pestles are interchangeable
TRUSTED_SUSTAINABILITY
Specifications
Size 20
Diameter (English) Shaft 0.15 in.
Diameter (Metric) Shaft 4mm
Capacity (English) 0.13 oz.
Capacity (Metric) 4 mL
Product Type Pestle and Graduated Tube
Graduations 0.5 to 4mL
Length (English) Pestle 8.46 in.
Length (Metric) Pestle 215mm
Length (English) Tube 4.9 in.
Length (Metric) Tube 125mm
Capacity 0.13 oz., 4mL