missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kontes™ Duall™ Tissue Grinders

Combined conical and cylindrical surfaces improve grinding efficiency

Brand:  DWK Life Sciences 885450-0025

 View more versions of this product

Product Code. 10707223

  • € 257.00 / Each
Estimated Shipment: 19-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Combined conical and cylindrical surfaces improve grinding efficiency

  • Can be motor driven
  • Choose from all-ground glass style or smooth glass tube with PTFE pestle threaded to stainless-steel shaft
  • Interchangeable tubes and pestles have 0.004 to 0.006 in. (0.10 to 0.15mm) clearance

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1.96 oz.
1.96 oz., 50mL
16mm
Ground Glass pestle and tube
8.85 in.
225mm
1.49 in.
50 mL
0.004 to 0.006 in.
0.62 in.
Ideal for connective tissue such as muscle, heart and lung
13.5 in.
345mm
25
38mm
0.1 to 0.15mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.