Learn More
DWK Life Sciences Kimble™ Kontes™ Duall™ Tissue Grinders with Safety-coated Tube
Click to view available options
Capacity:
0.1 oz., 3mL
0.16 oz., 5mL
0.59 oz., 15mL
Unit Size:
Each
Description
Autoclavable transparent coating
- With glass pestle
Specifications
Specifications
| Capacity (English) | 0.16 oz. |
| Clearance (English) | 0.004 to 0.006 in. |
| Clearance (Metric) | 0.1 to 0.15mm |
| Diameter (Metric) Shaft | 8mm |
| Diameter (English) Shaft | 0.31 in. |
| Capacity (Metric) | 5 mL |
| Length (English) Shaft | 8.66 in. |
| Length (English) Tube | 5.9 in. |
| Length (Metric) Shaft | 220mm |
| Length (Metric) Tube | 150mm |
| Show More |