Learn More
DWK Life Sciences KIMBLE™ EPA Water Analysis Vial, Without Closure
Click to view available options
Volume (Metric):
20 mL
30 mL
40 mL
60 mL
Color:
Clear
No. per Pack:
144
Unit Size:
Pack of 144
Pack of 432
Description
- Clear EPA vials are Converted from Type I, 33 expansion borosilicate glass tubing and amber EPA vials are converted from Type I, 51 expansion borosilicate glass tubing
- While vials are not pre-cleaned, they are packaged in a plastic shrink wrap module, lehr-clean, as they come from the end of the production lehr and then are placed into a corrugated shelf-pack
- Open-top closures and PTFE-faced silicone septa or solid-top PFTE-faced white rubber-lined closures are available separately
Compliance
Vials comply with guidelines for establishing test procedures for analyzing pollutants in water (Ref. U.S. EPA Methods 601, 602, 603, and 604).
Specifications
Specifications
| Diameter (English) Outer | 1.1 in. |
| Color | Clear |
| Diameter (Metric) Outer | 28 mm |
| Material | Borosilicate Glass |
| Volume (Metric) | 30 mL |
| For Use With (Application) | For water analysis |
| Height (English) | 2.75 in. |
| No. per Pack | 144 |
| Volume (English) | 1.01 oz. |
| Volume (Metric) Total | 30 mL |
| Show More |