Learn More
DWK Life Sciences Kimble™ KIMAX™ Conical-Bottom Glass Centrifuge Tubes: 15 mL Capacity
Product Code. 11825801
Click to view available options
Disposable:
Reusable
Single-use
Graduations:
0.1 mL
Permanent Red, 0.1 mL
Length (Metric):
118 mm
126 mm
130 mm
Unit Size:
Pack of 12
Pack of 125
Product Code. 11825801
Brand: DWK Life Sciences 4516415
Only null left
Add to basket
Add to basket
Withstand centrifugation up to 2980 RCF
  • With conical shaped bottom
  • Tubes are made from borosilicate glass in compliance with ASTM Specification E438, Type I, Class A or B glass requirements
TRUSTED_SUSTAINABILITY
Specifications
Calibration To contain
Certifications/Compliance USP, ASTM E 438
Capacity (Metric) 15 mL
Disposable Reusable
Material Borosilicate Glass
Tolerance 0 to 0.2mL in ±0.05mL, >0.2 to 1mL in ±0.1mL, >1 to 3mL in ±0.15mL, >3mL in ±0.2mL
Shape Conical
Length (Metric) 118 mm
Bottom Shape Conical
Closure Type Not Included
Writing Area Marking Spot
Diameter (Metric) 17 mm
Graduations Permanent Red, 0.1 mL
Product Type Centrifuge Tube
Max. RCF 2980 x g