Learn More
DWK Life Sciences Kimble™ Open-Top PolypropyleneScrew Thread Closures with Red PTFE-Faced Silicone Septa
Product Code. 11813033
missing translation for 'orderingAttributeHoverText'
Color:
Black
Blue
White
Pre-slit:
No
Yes
Closure Size:
13-425
8-425
9-425
missing translation for 'unitSize'
Pack of 1000
Product Code. 11813033
missing translation for 'mfr': DWK Life Sciences 73812BK9425
Only null left
Add to basket
Add to basket
Autoclavable and used for multiple injections
  • Manufactured using the highest quality raw materials
  • Designed for an ideal fit with Kimble Chase autosampler vials
  • Convenient assembly of caps and liners
  • Available with standard or pre-slit septa
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Material Polypropylene
Pre-slit No
Autoclavable Autoclavable
Closure Size 9-425