missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kontes™ Microscale Organic Chemistry LABSETS™ and Kits Components

Product Code. 10636285
Click to view available options
Product Type:
Disc
Distillation Column
Filter Adapter
Filter Flask
Hirsch Funnel
Long Neck Flask
Microflex Organic Chemistry Kit Connector
Microspatula
Short Neck Flask
thermometer Adapter
Quantity:
10/Pk.
100/Pk.
5/Pk.
Ea.
Unit Size:
Each
Pack of 10
Pack of 100
Pack of 5
This item is not returnable. View return policy

Product Code. 10636285

Brand: DWK Life Sciences 7480121000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Components for microscale organic chemistry LABSETS

  • Microflex Kits feature special chaste connectors and specially tooled glassware components, providing great versatility
  • They fit easily into lab bench drawers

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Microscale organic chemistry
For Use With (Equipment) Component part of Williamson standard organic chemistry labsets and kits
Includes Disc
Quantity 5/Pk.
Standard Taper Joint 14/20
Product Type Hirsch Funnel
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.