Learn More
DWK Life Sciences Kimble™ Kontes™ Microscale Organic Chemistry LABSETS™ and Kits Components
Product Code. 10267774
Click to view available options
Product Type:
Disc
Distillation Column
Filter Adapter
Filter Flask
Hirsch Funnel
Long Neck Flask
Microflex Organic Chemistry Kit Connector
Microspatula
Short Neck Flask
thermometer Adapter
Quantity:
10/Pk.
100/Pk.
5/Pk.
Ea.
Unit Size:
Each
Pack of 10
Pack of 100
Pack of 5
Product Code. 10267774
Brand: DWK Life Sciences 7480140000
Only null left
Add to basket
Add to basket
Components for microscale organic chemistry LABSETS
  • Microflex Kits feature special chaste connectors and specially tooled glassware components, providing great versatility
  • They fit easily into lab bench drawers
TRUSTED_SUSTAINABILITY
Specifications
Dimensions (L x W) 7.625 x 0.31 in. (193.6 x 8 mm)
For Use With (Application) Microscale organic chemistry
For Use With (Equipment) Component part of Williamson standard organic chemistry labsets and kits
Material Nickel-Stainless
Quantity 10/Pk.
Length (English) 7.625 in.
Length (Metric) 193.6 mm
Width (English) 0.31 in.
Width (Metric) 8 mm
Product Type Microspatula