Learn More
DWK Life Sciences Kimble™ Kontes™ Antibiotic Moisture Content Flasks
Product Code. 11858891
Click to view available options
:
Antibiotic moisture content flask; 2.2mL
Product Code. 11858891
Brand: DWK Life Sciences 8817000000
Only null left
Add to basket
Add to basket
For determining moisture loss on drying in tablets, troches, or capsules
Sample is weighed, transferred to the flask, and placed in a vacuum oven. Drying is done in the oven at a temperature of 60°C and a pressure of 5mm or less of mercury for three hours. The percent of loss is then calculated.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 2.2 mL
Material Borosilicate Glass
For Use With (Application) For determining moisture loss on drying in tablets, troches, or capsules
Standard Taper Joint 10/10
Bottom Shape Flat