Learn More
DWK Life Sciences Kimble™ Clear Screw Thread Sample Vials without Closures
Packaged in corrugated trays with partitions
Specifications
Color | Clear |
---|---|
Material | Borosilicate Glass |
Thread Style | Screw |
Certifications/Compliance | ASTM Type I, Class A |
Product Code | Brand | Volume (Metric) | Diameter (Metric) Outer | Height (Metric) | Closure Size | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Diameter (Metric) Outer | Height (Metric) | Closure Size | Price | Quantity & Availability | ||
11842983
|
DWK Life Sciences
608121528 |
2 mL | 15 mm | 28 mm | 13-425 |
N/A
|
N/A | ||
11822993
|
DWK Life Sciences
60812-2857 |
20 mL | 28 mm | 57 mm | 24-400 |
N/A
|
N/A | ||
11882983
|
DWK Life Sciences
608121940 |
6 mL | 19 mm | 40 mm | 15-425 |
N/A
|
N/A | ||
11832993
|
DWK Life Sciences
60812-2870 |
25 mL | 28 mm | 70 mm | 24-400 |
N/A
|
N/A | ||
11842993
|
DWK Life Sciences
608122895 |
40 mL | 28 mm | 95 mm | 24-400 |
N/A
|
N/A | ||
11862983
|
DWK Life Sciences
608121738 |
4 mL | 17 mm | 38 mm | 15-425 |
N/A
|
N/A | ||
11822983
|
DWK Life Sciences
608121232 |
1.8 mL | 12 mm | 32 mm | 8-425 |
N/A
|
N/A | ||
11852983
|
DWK Life Sciences
608121545 |
4 mL | 15 mm | 45 mm | 13-425 |
N/A
|
N/A | ||
11812993
|
DWK Life Sciences
608122385 |
24 mL | 23 mm | 85 mm | 20-400 |
N/A
|
N/A
|
||
11892983
|
DWK Life Sciences
608121965 |
12 mL | 19 mm | 65 mm | 15-425 |
N/A
|
N/A | ||
11802993
|
DWK Life Sciences
60812-2170 |
16 mL | 21 mm | 70 mm | 18-400 |
N/A
|
N/A | ||
11832983
|
DWK Life Sciences
60812-1235 |
2 mL | 12 mm | 35 mm | 8-425 |
N/A
|
N/A | ||
11872983
|
DWK Life Sciences
608121760 |
8 mL | 17 mm | 60 mm | 15-425 |
N/A
|
N/A | ||
Description
- Manufactured from clear 33 expansion borosilicate glass
Specifications
Clear | |
Borosilicate Glass | |
Screw | |
ASTM Type I, Class A |