Learn More
DWK Life Sciences Kimble™ Clear Screw Thread Sample Vials with Unattached PTFE-Faced/White Rubber Lined Closures
Product Code. 11882993
Click to view available options
Volume (Metric):
2 mL
4 mL
8 mL
12 mL
16 mL
24 mL
Height (Metric):
32 mm
45 mm
60 mm
65 mm
70 mm
85 mm
Diameter (Metric) Outer:
12 mm
15 mm
17 mm
19 mm
21 mm
23 mm
Closure Size:
13-425
15-425
18-400
20-400
8-425
Unit Size:
Pack of 200
Product Code. 11882993
Brand: DWK Life Sciences 60812B2
Only null left
Add to basket
Add to basket
Unattached closed top black phenolic closures
  • Manufactured from clear 33 expansion borosilicate glass
  • Closures includes PTFE-faced/white rubber liners
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Black
Classification A
Material (Liner) PTFE/Rubber
Color Clear
Diameter (Metric) Outer 17 mm
Material Borosilicate Glass
Volume (Metric) 8 mL
Thread Style Screw
Volume (English) 2.16 dr.
Closure Material Phenolic
Closure Type Screw Cap
Height (Metric) 60 mm
Certifications/Compliance ASTM Type I
Closure Size 15-425