Learn More
DWK Life Sciences Kimble™ Clear Screw Thread Sample Vials with Unattached PTFE-Faced/White Rubber Lined Closures
Click to view available options
Volume (Metric):
2 mL
4 mL
8 mL
12 mL
16 mL
24 mL
Height (Metric):
32 mm
45 mm
60 mm
65 mm
70 mm
85 mm
Diameter (Metric) Outer:
12 mm
15 mm
17 mm
19 mm
21 mm
23 mm
Closure Size:
13-425
15-425
18-400
20-400
8-425
Unit Size:
Pack of 200
Description
- Manufactured from clear 33 expansion borosilicate glass
- Closures includes PTFE-faced/white rubber liners
Specifications
Specifications
| Closure Color | Black |
| Classification | A |
| Material (Liner) | PTFE/Rubber |
| Color | Clear |
| Diameter (Metric) Outer | 21 mm |
| Material | Borosilicate Glass |
| Volume (Metric) | 16 mL |
| Thread Style | Screw |
| Volume (English) | 4.32 dr. |
| Closure Material | Phenolic |
| Show More |