missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Black Phenolic Screw-Thread Closures

Product Code. 15760409
Click to view available options
Neck Finish:
13-415
15-415
22-400
Unit Size:
Pack of 125
Pack of 144
Pack of 500
This item is not returnable. View return policy

Product Code. 15760409

Brand: DWK Life Sciences 7380222400

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly resistant to temperature and steam during autoclaving

  • Cemented-in PTFE-faced 14B white rubber liner
  • Three screw-thread finishes

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Material Phenolic Resin
No. per Case 144 Cs.
Neck Finish 22-400
Product Type Screw Cap
For Use With (Equipment) Bottle
Closure Liner PTFE/14B Rubber
Autoclavable Autoclavable
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.