missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Black Phenolic Screw-Thread Closures

Product Code. 10144804
Click to view available options
Neck Finish:
13-415
15-415
22-400
Unit Size:
Pack of 125
Pack of 144
Pack of 500
This item is not returnable. View return policy

Product Code. 10144804

Brand: DWK Life Sciences 7380215415

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Highly resistant to temperature and steam during autoclaving

  • Cemented-in PTFE-faced 14B white rubber liner
  • Three screw-thread finishes

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Material Phenolic Resin
Liner Material PTFE-faced Rubber
Neck Finish 15-415
Closure Included Yes
Product Type Screw Cap
For Use With (Equipment) Bottle
Autoclavable Autoclavable
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.