Learn More
DWK Life Sciences Kimble™ Kim-Kap™ Closures
Available in natural (clear) or choice of four colors
Brand: DWK Life Sciences 73662-18
Description
Available in natural (clear) or choice of four colors
- For plain-end culture tubes
- Keep tube contents sterile without need for cotton plugging
- Polypropylene
- Autoclavable
Specifications
Push Cap | |
Polypropylene | |
Non-sterile | |
Reusable |
Autoclavable | |
Red | |
18 mm Plain-end Culture Tubes |