Learn More
DWK Life Sciences™ KECK™ Clip for Spherical Joints, POM, S
Shop All DWK Life Sciences Products
Click to view available options
Color:
Dark Blue
Dark Red
Light Blue
Orange
Purple
Unit Size:
Pack of 100
Description
KECK™ spherical ground joint clamps represent a favourable alternative to conventional spherical ground joint clamps made of metal. They are easy to put on and take off. The good spring properties of the POM material prevent scratching of the glass surface. An important contribution to safety in the laboratory. The different colours make the sizes easy to distinguish from each other.
- Excellent dimensional stability and good spring properties
- No damage to the glass surface during attachment unlike conventional wire clamps
- Easy, safe and quick to put on and take off
- Colour coding per size
- Cost-effective alternative to conventional spherical ground joint clips made of metal
Specifications
Specifications
| Quantity | 100 |
| Material | POM |
| For Use With (Equipment) | Securing of Spherical Ground Joints |
| For Use With (Application) | Securing of spherical ground joints |
| Color | Dark Blue |