Learn More
DWK Life Sciences™ KECK™ Clip Assortment, for conical joints, from stainless steel (1.4310, blank)
Shop All DWK Life Sciences Products
Click to view available options
Size:
NS 10
NS 12
NS 14
NS 19
NS 24
NS 29
NS 34
NS 7
Unit Size:
Pack of 10
Description
KECK™ clamps are ground joint clamps used to secure standard ground joint connections in glass apparatus and are characterized by their easy handling and excellent resilience. The clamps are available in different materials (metal or polyoxymethylene).
- Dimensionally stable with particularly good spring properties
- Easy, safe and quick to set up
Specifications
Specifications
| Quantity | 10 |
| Material | Stainless Steel (1.4310, Blank) |
| For Use With (Equipment) | Secure Glass-to-Glass Joints |
| For Use With (Application) | Secure glass-to-glass joints |
| Size | NS 19 |