Learn More
KD Scientific Legato 380 Emulsifier Systems
Based on the Legato™ Series of syringe pumps. Utilizes a modified Legato™ 210P and Legato™ 270P
Brand:  KD Scientific 788380
Product Code. 13496028
  • € 12790.00 / Each
Estimated Shipment: 01-04-2025
to see stock.
Explore more special offers
Description
  • All the standard features of the Legato™ products still available
  • Custom emulsification program added to the software, only available with the Legato™ 380 system
  • Selectable emulsifying rates & volumes
  • Adjustable duration
Specifications
Specifications
100/240 V
35 kg (75 lb) @ 100% Force Selection
TFT
2
5 to 220.97 mL/min.
Legato 380 Emulsifier System
±0.35%