Learn More
Poeppelmann™ Kapsto™ Tapered Universal Protection Plugs, GPN600 Series
Manufactured from polyethylene. Poppelmann™ Kapsto™ Tapered Universal Protection Plugs, GPN600 Series can be used as plugs in bores or internal thread, or as caps for external thread.
Brand:  Poeppelmann™ 60004390000
 View more versions of this product
Product Code. 10033624
  • € 33.53 / Pack of 100
Estimated Shipment: 16-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Polyethylene
  • Natural color
Specifications
Specifications
Natural
42.5 mm
51.5 mm
Cone Closures
Use as plugs in bores or internal thread, or as caps for external thread
100 Pack
Polyethylene
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Poeppelmann™ Kapsto™ Tapered Universal Protection Plugs, GPN600 Series > Interior Dimensions: 43.9 dia. x 13.5mmH; External Dimensions: 40.2 dia. x 13.6mmH