Learn More
Poeppelmann™ KAPSTO™ GPN 610 Universal Protection Caps/Plugs
Excellent for closing off bore holes and internal threads, or to protect studs and external threads. Poeppelmann™ KAPSTO™ GPN 610 Universal Protection Caps/Plugs are manufactured from yellow PE-LD plastic.
Brand:  Poeppelmann™ 61000350000
 View more versions of this product
Product Code. 10229954
  • € 62.95 / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Universal use as a plug or cap
  • Acc. with VDA Band 19 and ISO 16232
  • Tapered design ideal for accomodating tolerances
Specifications
Specifications
Sealing Plug
45.2 mm
25 mm
100 Pack
55 mm
Polyethylene
Can be used universally as plug or cap. The tapered design is ideal for accommodating tolerances
Yellow
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers