missing translation for 'onlineSavingsMsg'
Learn More

Poeppelmann™ Kapsto™ Polyethylene Flathead Closing Plugs, GPN300 Series

Manufactured from polyethylene. Poppelmann™ Kapsto™ Polyethylene Flathead Closing Plugs, GPN300 Series protect internal threads and close off bores.

€ 17.15 - € 39.70

Specifications

Color Natural
Material Polyethylene
Type Closing Plug
For Use With (Application) For protection of internal threads and for closing off bores
Quantity 100 Pack
View More Specs

Products 3
Product Code Brand Size (Metric) Length (Metric) Price Quantity & Availability  
Product Code Brand Size (Metric) Length (Metric) Price Quantity & Availability  
10472355
View Documents
Poeppelmann™
30000190000
25 mm 10.5 mm
€ 17.60
Pack of 100
Estimated Shipment: 14-02-2025
to see stock.
Add to basket
 
10597194
View Documents
Poeppelmann™
30002320000
28 mm 9.5 mm
€ 17.15
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
10329204
View Documents
Poeppelmann™
30003110000
45 mm 18 mm
€ 39.70
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

  • Polyethylene
  • Natural color
Specifications

Specifications

Natural
Closing Plug
100 Pack
Polyethylene
For protection of internal threads and for closing off bores
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.