Learn More
Poeppelmann™ KAPSTO™ GPN 300 F Closures
Offering multifunctional use for plugging internal threads and bore holes. Poeppelmann™ KAPSTO™ GPN 300 F Closures are manufactured from natural-colored PE-LD plastic.
Brand: Poeppelmann™ 30100090000
This item is not returnable.
View return policy
Description
- Flat-headed design
- Firm fit due to crowned shank
- Production in acc. to VDA Band 19 and ISO 16232
Specifications
12.3 mm | |
Polyethylene | |
For internal threads and bores | |
100 |
Flat Top Closure Plug | |
Natural | |
8.9 mm | |
12.3 mm |