Learn More
K-425 Model SpeedDigestor
Set new standards in terms of digestion speed, flexible use and result reproducibility with the IR SpeedDigester. BUCHI K-425 Model SpeedDigestors provide increased sample throughput for nitrogen determination by Kjeldahl digestion and multiply instrument functionality when needs change.
Brand:  Buchi 1154252200
Product Code. 12048139
  • € 6025.00 / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Description
  • Fast heat transfer of infrared heaters to samples
  • Large time savings due to fast heating and cooling
  • Increased sample throughput due to shortened process time
  • Accelerated digestion step with continuous addition of H2O2
  • One instrument for both Kjeldahl and reflux digestion
  • All BUCHI sample tubes to be used (100mL, 300mL, 500mL)
  • Sample tubes compliant with ISO 6060 for COD or other reflux digestion (e.g. aqua regia)
  • Dedicated suction module for aqueous samples
  • Sealed suction module for effective transfer of harmful fumes
  • Effective neutralization of fumes by the Scrubber K-415
  • Safe operation and extended lifetime of fume hood
Specifications
Specifications
Speed Digestor
220/240 V ±10 V
620 mm
Feed - Kjeldahl, Chemicals/Agricultural, Beverage, Pharmaceutical / Cosmetics , Food , Environmental Analysis
310 mm
540 mm
50/60 Hz
1280 W
220/240 V ±10 V, 50/60 Hz, 1280 W
300mL Sample tubes and suction module with condensate trap
1 Ea.