missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ HPLC Remote Wall Bracket Kit
Brand: Justrite™ 28150
This item is not returnable.
View return policy
Specifications
Remote Wall Bracket Kit | |
Ideal for applications using the HPLC coalescing/Carbon filter | |
Steel | |
5 in. |
Polished Steel | |
HPLC Waste Collection Vessels | |
Polypropylene connector, female quick disconnect fitting, 5 in. tall no. 304 Stainless Steel holder and 2 self-tapping mounting screws | |
12.7 cm |
JTT001 International Catalogue May 2010
File size: 6.4 MB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Justrite™ HPLC Remote Wall Bracket Kit > Remote Wall Bracket Kit
Spot an opportunity for improvement?Share a Content Correction