Learn More
Tocris Bioscience™ JNJ 1013
Interleukin 1 Receptor Associated Kinase 1 (IRAK1) PROTAC™
Brand:  Tocris Bioscience™ 8029/5
226.65 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18084262
  • € 277.00 / 5mg
Explore more special offers
Description
Description
JNJ 1013 is a potent interleukin 1 receptor associated kinase 1 (IRAK1) PROTAC™ Degrader (DC50 = 3 nM; Dmax = 96%). In ABC DLBCL cell lines with MyD88 L265P mutation, JNJ 1013 potently inhibits IRAK1 downstream signaling pathways, induces apoptosis, and shows strong anti-proliferative effects. PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
N-(4-(4-(2-(((S)-1-((2S,4R)-4-Hydroxy-2-(((S)-1-(4-(4-methylthiazol-5-yl)phenyl)ethyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)amino)-2-oxoethoxy)piperidin-1-yl)-2-methoxyphenyl)-6-(1H-pyrazol-5-yl)picolinamide
5 mg
C46H55N9O7S
2597343-08-1
Active Degraders
98%
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers