Learn More
Cole-Parmer™ Jenway™ Glass Cuvettes
Brand:  Cole-Parmer™ 99610-30
102.60 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
 View more versions of this product
Product Code. 13197124
  • € 108.00 / Each
In Stock
to see stock.
Explore more special offers
Specifications
Cuvette
2 to 3.5 mL
Glass
10 mm
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer™ Jenway™ Glass Cuvettes > Capacity: 2 to 3.5mL