Learn More
Cole-Parmer™ JD Honigberg™ Ice-Pak Cold Packs
Brand:  Cole-Parmer™ 06345-40
Product Code. 11721029
Specifications
12 Pk.
Refrigerate Samples For Shipping
-6°C
Cold Pack
266.7 x 203.2 x 38.1 mm
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer™ JD Honigberg™ Ice-Pak Cold Packs > Dimensions (L x W x H): 266.7 x 203.2 x 38.1mm