Learn More
GPE Scientific™ J Young Finger Schlenk Tube with Fast Thread Tap
GPE Scientific™ J Young Finger Schlenk Tube with Fast Thread Tap is specially designed for use with Schlenk lines and vacuum manifolds.
Brand: GPE Scientific™ JFF/5
This item is not returnable.
View return policy
Description
- Solidly constructed for use with vacuum manifolds
- Designed for use with Schlenk lines
- Includes fast thread tap
Specifications
Includes fast thread tap | |
Schlenk lines and vacuum manifolds | |
Finger Schlenk tube | |
5 mL |
Product Content Correction
Product Title
GPE Scientific™ J Young Finger Schlenk Tube with Fast Thread Tap > Capacity: 5mL