missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isoxazole-5-carbonyl chloride, TECH, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Specifications
Specifications
CAS Min % | 89.5 |
CAS Max % | 100.0 |
Assay Percent Range | 90% |
Quantity | 10 g |
Formula Weight | 131.52 |
Percent Purity | 90% |
Chemical Name or Material | Isoxazole-5-carbonyl chloride |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Isoxazole-5-carbonyl chloride, TECH, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction