Learn More
Brady™ ISO Square Polyester Safety Sign 'Fire Alarm Flashing Light'
ISO Safety Sign displaying flashing fire alarm and flames.
Brand: Brady™ 311217
Description
- To indicate a device that flashes to indicate that the fire alarm has operated
Specifications
Class C | |
Gloss | |
1 Sign | |
Surface Mount | |
3.94 in. | |
-40°C to +66°C | |
-40°F to +150°F | |
100 x 100 mm | |
Safety Sign |
F/F018/NT/PE-PHOLUMC-100X100-1 | |
Polyester | |
Permanent | |
Facility Identification, Safety Identification | |
3.94 in. | |
Square | |
Universal | |
Fire Alarm Flashing Light | |
Photoluminescent on Red |