Learn More
Brady™ ISO Square Polyester Safety Sign 'Fire Alarm Flashing Light'
ISO Safety Sign displaying flashing fire alarm and flames.
Brand:  Brady™ 311219
 View more versions of this product
Product Code. 17851775
  • € 9.30 / Each
Estimated Shipment: 02-05-2025
to see stock.
Explore more special offers
Description
  • To indicate a device that flashes to indicate that the fire alarm has operated
Specifications
Specifications
Class C
Gloss
1 Sign
Surface Mount
7.87 in.
-40°C to +66°C
-40°F to +150°F
200 x 200 mm
Safety Sign
F/F018/NT/PE-PHOLUMC-200X200-1
Polyester
Permanent
Facility Identification, Safety Identification
7.87 in.
Square
Universal
Fire Alarm Flashing Light
Photoluminescent on Red
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Brady™ ISO Square Polyester Safety Sign 'Fire Alarm Flashing Light' > ISO Safety Sign Fire Alarm; Color: Photoluminescent on Red; Dimensions (L x W): 200 x 200 mm; Certifications/Compliance: Class C