missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Iso-Sensitest Agar + Sheep Blood

Product Code. 10211123
Click to view available options
Quantity:
10 Plates
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 10211123

Brand: Thermo Scientific™ PB5003A

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Support accurate antimicrobial susceptibility testing with this Agar, offering optimal conditions for a wide range of microorganisms.

Support accurate antimicrobial susceptibility testing with ISO Sensitest Agar enriched with sheep blood, offering optimal conditions for a wide range of microorganisms.

TRUSTED_SUSTAINABILITY

Specifications

Color Red
Content And Storage 2°C to 12°C
Format 90 mm Plate
Manufacturing Origin Germany
Product Type Agar Plate
Fill Weight 19 g
Quantity 10 Plates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.