Learn More
ISE Buffer Ammonia, Reagecon™
Shop All Reagecon Products
Click to view available options
Quantity:
500 mL
Specifications
Specifications
Quantity | 500 mL |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
ISENH4524K1 | Certificate of Analysis | 25/02/2025 | ISENH45 |
ISENH4524E1 | Certificate of Analysis | 27/08/2024 | ISENH45 |
ISENH4523F1 | Certificate of Analysis | 25/07/2024 | ISENH45 |
ISENH4521M1 | Certificate of Analysis | 18/10/2022 | ISENH45 |
Product Content Correction
Product Title
ISE Buffer Ammonia, Reagecon™ >