missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iridium for AAS 1g/L HCl matrix, Fisher Chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
€ 152.00 - € 525.00
Specifications
Concentration | 1g/L |
---|---|
Solution Contains | HCl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12993165
|
Fisher Chemical
B 027 100 |
100 mL |
€ 152.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12903175
|
Fisher Chemical
B 027 500 |
500 mL |
€ 525.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
1g/L | |
HCl |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Single-element Standard Solution for AAS. Iridium (Ir) 1000mg/l in HCl 10%
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501-Dispose of contents/container to .