Learn More
Masterflex™ IQ Masterflex Pumps
IQ Masterflex Pumps service provision verifies that a fluid handling pump is installed and operating correctly.
Brand: Masterflex™ 4100638
This item is not returnable.
View return policy
Description
Documented collection of activities necessary to:
- Estabilish if an instrument is delivered as designed and specified
- Determine if it is properly installed in the selected environment
- Ensuring that this environment is suitable for the instrument
Specifications
Masterflex™ Pumps | |
IQ |
Product Content Correction
Product Title
Masterflex™ IQ Masterflex Pumps > Assay Service Type: IQ