Learn More
Ipatasertib, MedChemExpress

MedChemExpress Ipatasertib (GDC-0068) is a highly selective and ATP-competitive pan-Akt inhibitor with IC50s of 5, 18 and 8 nM for Akt1, Akt2 and Akt3, respectively.

€ 108.00 - € 754.50
Specifications
Chemical Name or Material | Ipatasertib |
---|---|
Physical Form | Powder |
CAS | 1001264-89-6 |
Color | Off-White |
Purity Grade Notes | Research |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16486348
![]() |
Medchem Express
HY-15186 |
10 mg |
€ 166.50
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16478937
![]() |
Medchem Express
HY-15186 |
5 mg |
€ 108.00
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16422467
![]() |
Medchem Express
HY-15186 |
50 mg |
€ 498.00
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16486656
![]() |
Medchem Express
HY-15186 |
100 mg |
€ 754.50
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ipatasertib (GDC-0068) is a highly selective and ATP-competitive pan-Akt inhibitor with IC50s of 5, 18 and 8 nM for Akt1, Akt2 and Akt3, respectively.Specifications
Ipatasertib | |
1001264-89-6 | |
Research | |
Cancer-Kinase/protease | |
DMSO : 220 mg/mL (480.35 mM; Need ultrasonic) | |
458.0 | |
95.0% |
Powder | |
Off-White | |
C24H32ClN5O2 | |
GDC-0068 RG7440 | |
ClC1=CC=C([C@@H](CNC(C)C)C(N2CCN(C3=C([C@H](C)C[C@H]4O)C4=NC=N3)CC2)=O)C=C1 | |
458.0 | |
Research |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.