missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Platinum™ Pfx DNA Polymerase

Product Code. p-4926979
Click to view available options
Quantity:
100 Reactions
250 Reactions
500 Reactions
Unit Size:
100 reactions
250 reactions
500 reactions
This item is not returnable. View return policy

Product Code. 10532693

Brand: Invitrogen™ 11708013

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Platinum Pfx DNA Polymerase (100 units total, at 2.5U/μL) is a supplied with a vial (1mL) of 10X PCRx Enhancer solution, a vial (1mL) of 10X Pfx amplification buffer, and a vial (1mL) of 50mM MgSO4. Designed for 100 reactions (at 1 unit each in a 50μL reaction volume).

Platinum Pfx DNA Polymerase is ideal for amplification of DNA fragments for high-fidelity PCR applications.

Platinum Pfx DNA Polymerase is ideal for amplification of DNA fragments for high-fidelity PCR applications. High fidelity is provided by a proprietary enzyme preparation containing recombinant DNA polymerase from Thermococcusspecies KODwith proofreading (3'-5' exonuclease) activity. Platinumantibody technology provides a simple, automatic, hot-start method that improves PCR specificity. PCRx Enhancer Solution is included for higher primer specificity, broader magnesium concentration allowances, broader annealing temperature parameters, and improved thermostability of PlatinumPfxDNA Polymerase. The PCRx Enhancer Solution also helps optimize PCR of problematic and/or GC-rich templates.

• 26 times higher fidelity than Taq DNA polymerase
• Amplification of fragments up to 12 kb
• Room temperature reaction assembly

Applications
Amplification of DNA from complex genomic, viral, and plasmid templates; and RT-PCR. Platinum Pfx DNA Polymerase is especially effective when very high levels of fidelity are required.

Unit definition
One unit incorporates 10 nmoles of deoxyribonucleotide into acid-precipitable material in 30 minutes at 74°C.

Order Info

Approved for shipment on Wet or Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 Reactions
Includes Platinum Pfx DNA Polymerase (100 units total, at 2.5 U/μL) is a supplied with:
• 10X PCRx Enhancer solution, 1 mL
• 10X Pfx amplification buffer, 1 mL
• 50 mM MgSO4, 1 mL

Designed for 100 reactions (at 1 unit each in a 50 μL reaction volume)
Store at -20°C.

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.