Learn More
Memmert™ Interior Socket
Product Code. p-7220932 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP260eco
HPP410eco
HPP750eco
IPP1060eco
IPP110eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
Product Code. 17120285
Brand: Memmert™ IPP750ecoR3
Only null left
Add to basket
Add to basket
Interior IP68 socket (for temperature restriction +70°C option) 230V, 2. A. Cannot be switched individually, moisture tight IP68
  • Accessory
  • Ampacity 230 V/2.2 A
  • Can be switched off with the On/Off switch
Specifications
For Use With (Equipment) IPP750eco
Voltage 230 V