Learn More
WTW™ Interface Cable
Product Code. 10239060
Click to view available options
For Use With (Equipment):
PC to 4-pin socket
Unit Size:
Each
Product Code. 10239060
Brand: WTW™ 902841
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Type Interface Cable
For Use With (Application) For connecting an instrument with 4-pin socket to a PC via the RS-232 interface
For Use With (Equipment) PC to 4-pin socket