Learn More
Instrument Check Standard 8 For Environmental Analysis, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
€ 215.00 - € 346.00
Specifications
Chemical Name or Material | ICP Instrument Check Standard 8 |
---|---|
Packaging | Triple-leached LDPE bottle |
Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
Grade | Environmental |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15698235
|
SPEX Certiprep
CALMIX8-100 |
125 mL | Triple-leached LDPE bottle |
€ 215.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15608245
|
SPEX Certiprep
CALMIX8-500 |
500 mL | Triple-leached LDPE bottle |
€ 346.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Al | 50μg/mL |
As | 50μg/mL |
Co | 50μg/mL |
Cr | 50μg/mL |
Cu | 50μg/mL |
K | 50μg/mL |
Na | 50μg/mL |
P | 50μg/mL |
Pb | 50μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
ICP Instrument Check Standard 8 | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Environmental | |
2% HNO3 |
Triple-leached LDPE bottle | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
Each component at 50μg/mL in 2% HNO3: Aluminum Arsenic Chromium Cobalt Copper Lead Phosphorus Potassium Sodium |
Safety and Handling
Warning: Causes skin irritation, serious eye irritation Wear protective gloves/eye/face protection. If in eyes, rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If skin/eye irritation occurs, get medical advice/attention.
missing translation for 'dotInformation' : Hazardous
Recommended Storage : Ambient
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
- MIXTURE LIST-Contains : nitric acid
Product intended for laboratory use only