Learn More
Greiner Bio-One Insect (Drosophila) Breeding Conical Container
Ideally suited for the cultivation of Drosophila melanogaster.
Brand:  Greiner Bio-One 199101
 View more versions of this product
Product Code. 10517381
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Bottom part of container made of either polystyrene or polypropylene
  • Compatible with gas-permeable ceaprene stopper made of water-repellent material
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Conical
22 mm
63 mm
Drosophila Breeding Container
16 mL
Cultivation of Drosophila melanogaster
Polystyrene
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Greiner Bio-One Insect (Drosophila) Breeding Conical Container > Polystyrene; Volume: 16mL