missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ InPro™ pH Electrode

Low-maintenance pH probe. Mettler Toledo™ InPro™ pH Electrode complete with xerolyt extra solid polymer reference system, open junction, and built-in temperature sensor to allow reliable and accurate temperature-compensated pH measurement.

Brand:  Mettler Toledo™ 52000656

Product Code. 10677175

  • € 833.00 / Each
Estimated Shipment: 07-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Can be installed through bottom or floor of a vessel or tank.
  • Temperature range of 32-266°F.
  • Outstanding pressure resistance of up to six bar fully covers the pressure range normally encountered in processing.
  • Patented silver-ion barrier.
  • VarioPin connector in heavy-duty PEEK-based material. T
  • emperature range: 0° to 80°C.
  • pH range: 0 to 14.
  • Adapter cable sold separately.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

VarioPin
120 mm
0 to 14
InPro pH Electrode
PTFE
0°C to 80°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mettler Toledo™ InPro™ pH Electrode > Connector Types: VarioPin

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.