Learn More
Gosselin™ Inoculating T-Shaped Spreaders
For spreading over the surface of solid media in microbiological applications. Gosselin™ Inoculating T-Shaped Spreaders are manufactured from polystyrene plastic.
Brand: Gosselin™ ETAT-01
77.21 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Description
- More precise
- Sterile and disposable
- Packed in tamper-evident ZIP bags
Specifications
Single-use | |
148 mm | |
T-Shape |
Green | |
Pack of 500 | |
Sterile |