Learn More
Eppendorf™ Innova™ 44R Incubator Shaker
Use these stackable shakers for high speed applications and heavy workloads. New Brunswick Scientific™ Innova™ 44R Incubator Shakers provide broad temperature capabilities for culturing a wide variety of organisms in flasks up to 5L.
Brand: Eppendorf™ M1282-0016
20196.84 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
Description
- Cast iron triple-eccentric counterbalanced drive
- Refrigeration module is built into the Innova 44R, or can be easily added to 44, at any time
- Stackable – up to three units for maximum space savings
- Programmable Innova controls automatically changes temperature, speed, and optional photosynthetic and UV germicidal lights at timed intervals
- Versatile accessory platform (sold separately) accommodates flasks up to 5 L
- Slide-out platform mechanism – provides easy and effortless access to flasks located in the front and back of the shaker
- Built-in water reservoir humidifies chamber to reduce sample evaporation, while also protecting unit from spills, includes a drain for easy cleaning
- Pull-out service module allows access to all electronic and heating/cooling components, without having to unstack the units
Specifications
51 mm | |
230 V, 50 Hz | |
5 L | |
50 Hz | |
119.3 cm | |
Innova 44R |
25 to 300 rpm | |
230 V | |
Incubator Shaker | |
87.3 cm | |
135 cm | |
0.01 to 99:59 hr. |