Learn More
Vitlab™ Industrial Scoop
Use these Corrosion resistant scoops for applications in the food industry. VITLAB™ Industrial Scoops made of HDPE are almost unbreakable. Plastic scoops are lighter than metal, don’t dent and feature good chemical resistance.
Brand: Vitlab™ 40493
Description
- Clear HDPE scoop
- Conical in shape with tapered filling edge
Specifications
Industrial Scoop | |
150 mL | |
For contact with food |
HDPE | |
Natural | |
250 mm |